missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Fructose-6-phosphate disodium salt, 95%

Product Code. 15447619
Click to view available options
Quantity:
100 mg
500 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

D-Fructose-6-phosphate disodium salt is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-6-phosphate disodium salt is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).

Solubility
Soluble in water (100 mg/ml).

Notes
Store at -20°C. Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26177-86-6
Molecular Formula C6H13Na2O9P
Molecular Weight (g/mol) 306.11
MDL Number MFCD16661244
InChI Key VAPLWAFNCWDFQF-UHFFFAOYNA-N
PubChem CID 129893509
IUPAC Name sodium;[(2R,3R,4S)-2,3,4,6-tetrahydroxy-5-oxohexyl] dihydrogen phosphate
SMILES [Na].[Na].OCC(=O)C(O)C(O)C(O)COP(O)(O)=O

Specifications

Color White
Quantity 100 mg
Solubility Information Soluble in water (100mg/ml).
Formula Weight 304.1
Percent Purity 95%
Physical Form Powder
Chemical Name or Material D-Fructose-6-phosphate disodium salt
compliance-icons
Product Identifier
  • D-Fructose-6-phosphate disodium salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : D-Fructose-6-phosphate disodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.