missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Fructose, 99%

Product Code. 10634642 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10634642

Brand: Thermo Scientific Chemicals 161350025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D(-)-Fructose, 99%, C6H12O6, CAS Number-57-48-7, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, arabino-hexulose, d--fructose, d---fructose, d-levulose, fructose, d, furucton, keto-d-fructose, krystar 300, sugar, fruit, 500g, Passes test, 98.5% min, 31,321, CHEBI:48095, 50ppm max, White, 200-333-3 | CAS: 57-48-7 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 103.0°C to 105.0°C
Color White
Loss on Drying 0.5% (70°C, 4 hrs) (vacuum) max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 31,321
Merck Index 15,4302
Specific Rotation Condition − 92.25 (20.00°C c=10,H2O,on dry sub.)
Specific Rotation − 92.25
Solubility Information Solubility in water: 3750g/L (20°C). Other solubilities: soluble in alcohol,pyridine,ether,ethylamine,and methylamine
Formula Weight 180.16
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D(-)-Fructose
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.