missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(-)-Fructose, Certified AR for Analysis, Fisher Chemical™
59.35 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
1 kg
Description
D(-)-Fructose, Certified AR for Analysis
Specifications
Melting Point | 103°C |
Color | White |
pH | 5 to 7 |
Packaging | Plastic powder jar |
Quantity | 100 g |
Formula Weight | 180.16g/mol |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'