missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Fructose, specified according the requirements of USP, Thermo Scientific Chemicals

Product Code. 15209315
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 103.0°C to 105.0°C
Ignition Residue 0.5% max
Quantity 1 kg
Merck Index 15,4302
Specific Rotation − 92.25
Solubility Information Solubility in water: 3750g/L (20°C). Other solubilities: soluble in alcohol,pyridine,ether,ethylamine,and methylamine
Formula Weight 180.16
Chemical Name or Material D(-)-Fructose
Product Identifier
  • D(-)-Fructose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.