missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-(+)-Galactose, 98% Product Code.: 11475183

Thermo Scientific Chemicals D-(+)-Galactose, 98%

Product Code. 11475183
1000 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11475183

Brand: Thermo Scientific Chemicals A12813.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-(+)-Galactose, CAS # 59-23-4, is a monosaccharide that is an important constituent of glycolipids and glycoproteins. It is also used as an energy source by organisms.

Galactose is a simple monosaccharide used as a component of the galactosyltransferase labeling buffer. It serves as an energy source and an important constituent of glycolipids and glycoproteins. Since it is a component of antigens, it plays a vital role in the determination of blood type within the ABO blood group system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • D-(+)-Galactose is a monosaccharide used as a component of the galactosyltransferase labeling buffer. The deficiency of galactosyl-1-phosphate uridyltransferase causes an error in galactose metabolism known as galactosemia, increasing the galactose levels in the blood
  • D-(+)-Galactose is a component of the disaccharide lactose and is released upon hydrolysis by β-galactosidase enzymes

Application

  • This compound can be used as a source of energy as it is an essential constituent of glycolipids and glycoproteins
  • It is a component of antigens which makes it important for determining blood type within the ABO blood group system
  • It has been used to induce the expression of uncoupling protein (UCP) in yeast transformants
  • It is frequently used as a source of carbon in vitro culture media
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6H12O6
MDL Number MFCD00151230

Specifications

Melting Point 164°C to 168°C
Density 1.5
Quantity 1000 g
Beilstein 1724619
Merck Index 14,4335
Solubility Information Soluble in water.
Optical Rotation +80° (c=5 in Water, 24h)
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material D-(+)-Galactose

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM13L053 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog NumberA12813.0B,A12813.18,A12813.30
Lot Number10252954 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberA12813.0B,A12813.18,A12813.30
Lot NumberT23K020 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog NumberA12813.0B,A12813.18,A12813.30
Lot NumberQ23K011 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA12813.0B,A12813.18,A12813.30
Lot NumberN16K038 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog NumberA12813.0B,A12813.18,A12813.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-(+)-Galactose, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.