missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Galacturonic acid monohydrate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
D-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.
Solubility
Soluble in water
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
D-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.
Solubility
Soluble in water
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Quantity | 25 g |
Beilstein | 1727087 |
Merck Index | 14,4337 |
Solubility Information | Soluble in water |
Optical Rotation | +53° (c=10 in Water) |
Formula Weight | 212.15 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | D-Galacturonic acid monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'