missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Gluconic acid, 50% aq. soln., Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
2500 mL
Description
Gluconic acid occurs naturally in fruit, honey, and wine. As a food additive it is an acidity regulator. It is also used in cleaning products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Gluconic acid occurs naturally in fruit, honey, and wine. As a food additive it is an acidity regulator. It is also used in cleaning products.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Gluconic acid occurs naturally in fruit, honey, and wine. As a food additive it is an acidity regulator. It is also used in cleaning products.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
UN Number | UN3265 |
Beilstein | 1726055 |
Merck Index | 144456 |
Solubility Information | Soluble in water. |
Formula Weight | 196.16 (Anhydrous) |
Concentration or Composition (by Analyte or Components) | 50% aq. soln. |
Density | 1.234 |
Quantity | 500 mL |
Optical Rotation | +10° (Neat) |
Refractive Index | 1.418 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'