missing translation for 'onlineSavingsMsg'
Learn More

D-Gluconic acid, calcium salt, 99%

Product Code. 10358250 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 10358250

Brand: Thermo Scientific Acros 211060010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 299-28-5 | C12H22CaO14 | 430.372 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-28-5
Molecular Formula C12H22CaO14
Molecular Weight (g/mol) 430.372
MDL Number MFCD00064209
InChI Key NEEHYRZPVYRGPP-IYEMJOQQSA-L
Synonym calcium gluconate, calcium d-gluconate, calciofon, calglucon, glucobiogen, ebucin, calcicol, calcipur, calglucol, dragocal
PubChem CID 9290
IUPAC Name calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]

Specifications

Color White
Loss on Drying 2.0 % max. (1 g, 105°C, 16 hrs)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Plastic bottle
Linear Formula (HOCH2[CH(OH)]4CO2)2Ca
Quantity 1 kg
Beilstein 03, 544
Merck Index 15, 1671
Specific Rotation Condition + 10.20 (22.00°C c=1,H2O)
Specific Rotation + 10.20
Formula Weight 430.38
Percent Purity 0.99
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D-Gluconic acid, calcium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.