missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucono-1,5-lactone, 99%, Thermo Scientific Chemicals

Product Code. 11344529
Click to view available options
Quantity:
250 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Glucono delta-lactone (GDL) is a kind of excellent multifunctional food additive. It is mainly used as protein coagulant, acidifier, pickling, or leavening agent. expander, preservative, seasonings, sequestrant, chelating agent, color-preserver, etc. it is widely applied to food, daily chemical industry, pharmaceuticals, cosmetics. Plastic and resinous modification, electrode position and polish, metal-surface cleaning and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glucono delta-lactone (GDL) is a kind of excellent multifunctional food additive. It is mainly used as protein coagulant, acidifier, pickling, or leavening agent. expander, preservative, seasonings, sequestrant, chelating agent, color-preserver, etc. it is widely applied to food, daily chemical industry, pharmaceuticals, cosmetics. Plastic and resinous modification, electrode position and polish, metal-surface cleaning and organic synthesis.

Solubility
Soluble in water 500 g/L (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep awy from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-80-2
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00006647
InChI Key PHOQVHQSTUBQQK-UHFFFAOYNA-N
Synonym gluconolactone, delta-gluconolactone, d-glucono-1,5-lactone, d-gluconolactone, 1,5-gluconolactone, d-gluconic acid lactone, gluconic acid lactone, glucono delta-lactone, 1,5-d-gluconolactone, gluconic lactone
PubChem CID 7027
ChEBI CHEBI:16217
SMILES OCC1OC(=O)C(O)C(O)C1O

Specifications

Melting Point 154°C to 159°C (decomposition)
Quantity 250 g
Beilstein 83286
Merck Index 14,4457
Solubility Information Soluble in water 500g/L (20°C).
Optical Rotation +65° (c=4 in Water)
IUPAC Name (3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
Formula Weight 178.14
Percent Purity 99%
Chemical Name or Material D-(+)-Glucono-1,5-lactone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'