missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Glucose-6-Phosphate, Disodium Salt Hydrate, MP Biomedicals™
Click to view available options
Quantity:
500 mg
5 g
25 g
Description
Glucose 6-phosphate (also known as Robison ester) is glucose sugar phosphorylated on carbon 6. This compound is very common in cells, as the vast majority of glucose entering a cell will become phosphorylated in this way. Because of its prominent position in cellular chemistry, glucose 6-phosphate has many possible fates within the cell.
Glucose 6-phosphate lies at the start of two major metabolic pathways: Glycolysis and Pentose phosphate. In addition to these metabolic pathways, glucose 6-phosphate may also be converted to glycogen or starch for storage. This storage is in the liver and muscles in the form of glycogen for most multicellular animals, and in intracellular starch or glycogen granules for most other organisms.
Specifications
Specifications
pH | 7 to 9 (1% aq. soln.) |
Quantity | 5 g |
Solubility Information | Soluble in distilled water or dilute buffer. |
Optical Rotation | +30–38° (c=1, water) |
Formula Weight | 304.2 (anhydrous) |
Percent Purity | ≥98% |
Physical Form | Powder |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'