Learn More
Thermo Scientific Chemicals D-(+)-Glucose, anhydrous, 99%
Description
D-Glucose is used in various metabolic processes including enzymic synthesis of cyclohexyl-alpha and beta -D-glucosides. It is involved in the detection of type 2 diabetes mellitus and the most widely used aldohexose in living organisms. Its availability influences psychological processes since it is a primary energy source for the brain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
D-Glucose is used in various metabolic processes including enzymic synthesis of cyclohexyl-alpha and beta -D-glucosides. It is involved in the detection of type 2 diabetes mellitus and the most widely used aldohexose in living organisms. Its availability influences psychological processes since it is a primary energy source for the brain.
Solubility
Soluble in water, hot glacial acetic acid, pyridine, aniline and dimethyl sulfoxide. Slightly soluble in methanol, ethanol and acetone.
Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 147°C to 153°C |
Assay Percent Range | 99% |
Quantity | 500 g |
Beilstein | 1281608 |
Merck Index | 14,4459 |
Solubility Information | Soluble in water,hot glacial acetic acid,pyridine,aniline and dimethyl sulfoxide. Slightly soluble in methanol,ethanol and acetone. |
Formula Weight | 180.16 |
Percent Purity | 99% |
Chemical Name or Material | D-(+)-Glucose, anhydrous |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X25K053 | Certificate of Analysis | 08/11/2024 | A16828.0C,A16828.36,A16828.0E |
X09K038 | Certificate of Analysis | 15/10/2024 | A16828.0C,A16828.36,A16828.0E |
10252490 | Certificate of Analysis | 26/09/2024 | A16828.0C,A16828.36,A16828.0E |
R13K017 | Certificate of Analysis | 11/06/2024 | A16828.0C,A16828.36,A16828.0E |
P20K028 | Certificate of Analysis | 11/06/2024 | A16828.0C,A16828.36,A16828.0E |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.