missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(+)-Glucose Anhydrous, Extra Pure, SLR, Fisher Chemical™
£50.70 - £230.00
Chemical Identifiers
CAS | 50-99-7 |
---|---|
Molecular Formula | C6H12O6 |
Molecular Weight (g/mol) | 180.156 |
MDL Number | 148912 |
InChI Key | GZCGUPFRVQAUEE-SLPGGIOYSA-N |
Synonym | 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form |
PubChem CID | 107526 |
ChEBI | CHEBI:42758 |
IUPAC Name | (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
SMILES | C(C(C(C(C(C=O)O)O)O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10335850
|
Fisher Chemical
G/0450/53 |
500 g | Plastic powder jar |
£50.70
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10395890
|
Fisher Chemical
G/0450/60 |
1 kg | Plastic powder jar |
£68.20
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10254480
|
Fisher Chemical
G/0450/61 |
2 kg | Plastic powder jar |
£110.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10539380
|
Fisher Chemical
G/0450/65 |
5 kg | Plastic keg |
£230.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
D(+)-Glucose Anhydrous, Extra Pure, SLR
Chemical Identifiers
50-99-7 | |
180.156 | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
107526 | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
C6H12O6 | |
148912 | |
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
CHEBI:42758 | |
C(C(C(C(C(C=O)O)O)O)O)O |
Specifications
50-99-7 | |
White | |
C6H12O6 | |
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
C(C(C(C(C(C=O)O)O)O)O)O | |
180.156 | |
CHEBI:42758 | |
Solid |
146°C | |
6 to 7 | |
148912 | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
107526 | |
180.16g/mol |