missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(+)-Glucose monohydrate, 99+%, extra pure, Thermo Scientific Chemicals
41.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Ignition Residue | 0.1% max |
Assay Percent Range | 99% min. (ELSD) (HPLC) |
Packaging | Plastic Drum |
Quantity | 5 kg |
Solubility Information | Solubility in water: soluble. |
Formula Weight | 198.17 |
Percent Purity | 99+% |
Grade | Extra Pure |
Chemical Name or Material | D(+)-Glucose monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'