missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals β-D-Glucose pentaacetate, 98%

Product Code. 10495423
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-69-3
MDL Number MFCD00006597
InChI Key LPTITAGPBXDDGR-IBEHDNSVSA-N
Synonym beta-d-glucose pentaacetate, penta-o-acetyl-beta-d-glucopyranose, unii-v833p95coc, 1,2,3,4,6-penta-o-acetyl-beta-d-glucopyranose, 1,2,3,4,6-penta-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, 2s,3r,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, glucopyranose, pentaacetate, .beta.-d, penta-o-acetyl-β
PubChem CID 2724702
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C16H22O11
Quantity 5 g
Specific Rotation Condition + 5.00 (20.00°C c=1,CHCL3)
Specific Rotation +4° to +6° (20°C, 589nm) (c=1, CHCl3)
Molecular Weight (g/mol) 390.341
Formula Weight 390.33
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material β-D-Glucose pentaacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'