missing translation for 'onlineSavingsMsg'
Learn More

D-Glucuronic acid, 98%, Thermo Scientific Chemicals

Product Code. 10531984
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6556-12-3
Molecular Formula C6H10O7
Molecular Weight (g/mol) 194.14
MDL Number MFCD00064368
InChI Key IAJILQKETJEXLJ-QTBDOELSSA-N
IUPAC Name (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid

Specifications

Melting Point 158.0°C to 162.0°C
Color White
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Assay Percent Range 98%
Merck Index 15,4501
Synonym benzonitrile, 2,4,6-trichloro, 2,4,6-trichlorobenzenecarbonitrile, pubchem4822, acmc-1b2tu, pgodhcioipodfe-uhfffaoysa
SMILES O[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)C(O)=O
PubChem CID 522179
Formula Weight 194.14
Percent Purity 98%
Chemical Name or Material D-Glucuronic acid, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'