missing translation for 'onlineSavingsMsg'
Learn More

D-Glucuronic acid, 98+%

Product Code. 11363486
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11363486

Brand: Thermo Scientific Alfa Aesar L14350.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6556-12-3 | C6H10O7 | 194.14 g/mol

D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6556-12-3
Molecular Formula C6H10O7
Molecular Weight (g/mol) 194.14
MDL Number MFCD00077778
InChI Key IAJILQKETJEXLJ-QTBDOELSSA-N
Synonym 2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoic acid, aldehydo-d-glucuronic acid, unii-8a5d83q4rw, rel, d-glucuronic acid, d-+ glucuronic acid, d03jeu, d-glucuronic acid 5g
PubChem CID 65041
ChEBI CHEBI:47953
IUPAC Name (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
SMILES C(=O)C(C(C(C(C(=O)O)O)O)O)O

Specifications

Melting Point ∽158°C (decomposition)
Quantity 5 g
Beilstein 1727083
Merck Index 14,4465
Solubility Information Soluble in water.
Optical Rotation +35° (c=6 in Water)
Formula Weight 194.14
Percent Purity ≥98%
Chemical Name or Material D-Glucuronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.