missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Glucuronic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∽158°C (decomposition) |
Quantity | 25 g |
Beilstein | 1727083 |
Merck Index | 14,4465 |
Solubility Information | Soluble in water. |
Optical Rotation | +35° (c=6 in Water) |
Formula Weight | 194.14 |
Percent Purity | ≥98% |
Chemical Name or Material | D-Glucuronic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'