missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 6893-26-1 |
---|---|
Molecular Weight (g/mol) | 147.13 |
MDL Number | MFCD00063112 |
InChI Key | WHUUTDBJXJRKMK-GSVOUGTGSA-N |
Synonym | d-glutamic acid, h-d-glu-oh, d-glu, r-2-aminopentanedioic acid, 2r-2-aminopentanedioic acid, d-glutamate, d-2-aminopentanedioic acid, glutamic acid d-form, d--glutamic acid, d-glutaminic acid |
PubChem CID | 23327 |
ChEBI | CHEBI:15966 |
IUPAC Name | (2R)-2-aminopentanedioic acid |
SMILES | C(CC(=O)O)C(C(=O)O)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10571861
|
Thermo Scientific Acros
227890050 |
5 g | Glass bottle |
£33.67
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10154760
|
Thermo Scientific Acros
227890250 |
25 g | Glass bottle |
£112.56
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10482421
|
Thermo Scientific Acros
227891000 |
100 g | Glass bottle |
£323.61
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6893-26-1 | |
MFCD00063112 | |
d-glutamic acid, h-d-glu-oh, d-glu, r-2-aminopentanedioic acid, 2r-2-aminopentanedioic acid, d-glutamate, d-2-aminopentanedioic acid, glutamic acid d-form, d--glutamic acid, d-glutaminic acid | |
CHEBI:15966 | |
C(CC(=O)O)C(C(=O)O)N |
147.13 | |
WHUUTDBJXJRKMK-GSVOUGTGSA-N | |
23327 | |
(2R)-2-aminopentanedioic acid |
Specifications
6893-26-1 | |
White | |
Authentic | |
C5H9NO4 | |
MFCD00063112 | |
− 31.30 (20.00°C c=10,2 N HCl) | |
d-glutamic acid, h-d-glu-oh, d-glu, r-2-aminopentanedioic acid, 2r-2-aminopentanedioic acid, d-glutamate, d-2-aminopentanedioic acid, glutamic acid d-form, d--glutamic acid, d-glutaminic acid | |
WHUUTDBJXJRKMK-GSVOUGTGSA-N | |
(2R)-2-aminopentanedioic acid | |
23327 | |
147.13 | |
Powder |
200.0°C to 202.0°C | |
0.2% max. (105°C, 3 hrs) | |
99+% | |
HO2CCH2CH2CH(NH2)CO2H | |
15, 4505 | |
− 31.30 | |
Solubility in water: 7g/L (20°C). Other solubilities: practically insoluble in organic solvents | |
C(CC(=O)O)C(C(=O)O)N | |
147.13 | |
CHEBI:15966 | |
≥99% | |
D(-)-Glutamic acid, 99+% |
Safety and Handling
missing translation for 'einecsNumber' : 230-000-8
missing translation for 'tsca' : TSCA
Product Identifier
- D(-)-Glutamic acid
RUO – Research Use Only