Learn More
Thermo Scientific Chemicals D(-)-Glutamine 98%
£62.60 - £211.00
Chemical Identifiers
CAS | 5959-95-5 |
---|---|
Molecular Formula | C5H10N2O3 |
Molecular Weight (g/mol) | 146.146 |
MDL Number | MFCD00065607 |
InChI Key | ZDXPYRJPNDTMRX-GSVOUGTGSA-N |
Synonym | d-glutamine, h-d-gln-oh, d-2-aminoglutaramic acid, l +-glutamine, 2r-2-amino-4-carbamoylbutanoic acid, r-2,5-diamino-5-oxopentanoic acid, d-glutamic acid 5-amide, d-glutamin, unii-63hb36ca2y, d-gln |
PubChem CID | 145815 |
ChEBI | CHEBI:17061 |
IUPAC Name | (2R)-2,5-diamino-5-oxopentanoic acid |
SMILES | C(CC(=O)N)C(C(=O)O)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10068671
|
Thermo Scientific Chemicals
135160010 |
1 g | Glass bottle |
£62.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10271592
|
Thermo Scientific Chemicals
135160050 |
5 g | Glass bottle |
£211.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5959-95-5 | |
146.146 | |
ZDXPYRJPNDTMRX-GSVOUGTGSA-N | |
145815 | |
(2R)-2,5-diamino-5-oxopentanoic acid |
C5H10N2O3 | |
MFCD00065607 | |
d-glutamine, h-d-gln-oh, d-2-aminoglutaramic acid, l +-glutamine, 2r-2-amino-4-carbamoylbutanoic acid, r-2,5-diamino-5-oxopentanoic acid, d-glutamic acid 5-amide, d-glutamin, unii-63hb36ca2y, d-gln | |
CHEBI:17061 | |
C(CC(=O)N)C(C(=O)O)N |
Specifications
5959-95-5 | |
Authentic | |
C5H10N2O3 | |
MFCD00065607 | |
-30° to -34° (20°C, 589nm) (c=10, 2N HCl) | |
ZDXPYRJPNDTMRX-GSVOUGTGSA-N | |
(2R)-2,5-diamino-5-oxopentanoic acid | |
145815 | |
146.15 | |
Crystalline Powder |
White | |
98% | |
H2NCOCH2CH2CH(NH2)COOH | |
−32.00 (20.00°C 589nm,c=10,N HCl) | |
d-glutamine, h-d-gln-oh, d-2-aminoglutaramic acid, l +-glutamine, 2r-2-amino-4-carbamoylbutanoic acid, r-2,5-diamino-5-oxopentanoic acid, d-glutamic acid 5-amide, d-glutamin, unii-63hb36ca2y, d-gln | |
C(CC(=O)N)C(C(=O)O)N | |
146.146 | |
CHEBI:17061 | |
≥97.5% | |
D(-)-Glutamine, 98% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only