missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Leucine, 99%
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Arsenic (As) | 2ppm max. |
Color | White |
Heavy Metals (as Pb) | 10ppm max. |
Assay Percent Range | 99% |
Infrared Spectrum | Authentic |
Linear Formula | (CH3)2CHCH2CH(NH2)CO2H |
Iron (Fe) | 10ppm max. |
Beilstein | 04, 446 |
Loss on Drying | 0.2% max. |
Packaging | Glass Bottle |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'