missing translation for 'onlineSavingsMsg'
Learn More

BD D- Luciferin Potassium Salt

Synthetically made substrate which can be catalyzed by firefly luciferase

Brand:  BD 556877

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15858788

  • £705.00 / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

  • Also known as beetle luciferin
  • Lyophilized in form of monopotassium salt
  • Purified by HPLC to remove dehydroluciferin and L-Luciferin
  • Form dehydroluciferin in presence of ultraviolet light and/or moisture

Luminometer

Specifications

Specifications

Lyophilized
10 mg
Luminometer
D-Luciferin
-20°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers