missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Malic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
D-(+)-Malic acid used as an acidulant and flavoring agent, food additive. And it is also used in the place of the less sour citric acid in sour sweets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-(+)-Malic acid used as an acidulant and flavoring agent, food additive. And it is also used in the place of the less sour citric acid in sour sweets.
Solubility
Soluble in methanol, ethanol, acetone, ether.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
D-(+)-Malic acid used as an acidulant and flavoring agent, food additive. And it is also used in the place of the less sour citric acid in sour sweets.
Solubility
Soluble in methanol, ethanol, acetone, ether.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
Melting Point | 100°C to 104°C |
Density | 1.6 |
Quantity | 1 g |
Beilstein | 1723540 |
Merck Index | 14,5707 |
Solubility Information | Soluble in methanol,ethanol,acetone,ether. |
Optical Rotation | +28° (c=5.5 in pyridine) |
Formula Weight | 134.09 |
Percent Purity | ≥98% |
Chemical Name or Material | D- (+)-Malic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'