Learn More
D(+)-Malic acid, 98%, Thermo Scientific Chemicals
£19.80 - £182.91
Chemical Identifiers
CAS | 636-61-3 |
---|---|
Molecular Formula | C4H6O5 |
Molecular Weight (g/mol) | 134.09 |
MDL Number | MFCD00004245 |
InChI Key | BJEPYKJPYRNKOW-UHFFFAOYNA-N |
Synonym | d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate |
PubChem CID | 92824 |
ChEBI | CHEBI:30796 |
IUPAC Name | (2R)-2-hydroxybutanedioic acid |
SMILES | OC(CC(O)=O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10725653
|
Thermo Scientific Acros
153500010 |
1 g | Glass bottle |
£19.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10503131
|
Thermo Scientific Acros
153500050 |
5 g | Glass bottle |
£55.78
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10599243
|
Thermo Scientific Acros
153500250 |
25 g | Glass bottle |
£182.91
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
636-61-3 | |
134.09 | |
BJEPYKJPYRNKOW-UHFFFAOYNA-N | |
92824 | |
(2R)-2-hydroxybutanedioic acid |
C4H6O5 | |
MFCD00004245 | |
d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate | |
CHEBI:30796 | |
OC(CC(O)=O)C(O)=O |
Specifications
636-61-3 | |
White | |
C4H6O5 | |
MFCD00004245 | |
15, 5771 | |
Solubility in water: soluble. | |
OC(CC(O)=O)C(O)=O | |
134.09 | |
CHEBI:30796 | |
98% | |
Crystalline Powder |
99.0°C to 104.0°C | |
98% | |
HO2CCH2CH(OH)CO2H | |
03, 417 | |
d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate | |
BJEPYKJPYRNKOW-UHFFFAOYNA-N | |
(2R)-2-hydroxybutanedioic acid | |
92824 | |
134.09 | |
Authentic | |
D(+)-Malic acid |
Safety and Handling
GHS P Statement Causes serious eye irritation.
GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Danger
- D(+)-Malic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.