missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-Mannitol, 98+% Product Code.: 10021110

Thermo Scientific Chemicals D-Mannitol, 98+%

Product Code. 10021110
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10021110

Brand: Thermo Scientific Chemicals 125345000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 165.0°C to 170.0°C
Color White
Loss on Drying 0.3% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 01, IV, 2841
Merck Index 15, 5810
Specific Rotation +23° to +25° (20°C, 589nm) (PHEUR)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in pyridine and aniline,slightly soluble in lower alcohols,practically insoluble in most other common,organic solvents
IUPAC Name hexane-1,2,3,4,5,6-hexol
Formula Weight 182.17
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material D-Mannitol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469002 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number125340025,125340050,125345000,125340010,125341000
Lot NumberA0462460 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number125340025,125340050,125345000,125340010,125341000
Lot NumberA0457404 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number125340025,125340050,125345000,125340010,125341000
Lot NumberA0456649 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number125340025,125340050,125345000,125340010,125341000
Lot NumberA0456650 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number125340025,125340050,125345000,125340010,125341000
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    14

    Minutes

    56

    Seconds

    49

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-Mannitol, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.