missing translation for 'onlineSavingsMsg'
Learn More

D-Mannitol, ACS reagent, Thermo Scientific Chemicals

Product Code. 10358673
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 165.0°C to 170.0°C
Color White
Ignition Residue 0.01% max.
Loss on Drying 0.05% max. (5 g, 105°C, 2 hrs)
Infrared Spectrum Authentic
Packaging Plastic drum
Quantity 5 kg
Beilstein 01, IV, 2841
Merck Index 15, 5810
Specific Rotation +23.3° to +24.3° (25°C, 589nm) (Reagent ACS-directives)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in pyridine and aniline,slightly soluble in lower alcohols,practically insoluble in most other common,organic solvents
Formula Weight 182.17
Grade ACS Reagent
Insoluble Matter 0.01% max
Titratable Acid 0.0008 meq/g max
Physical Form Crystalline Powder
Chemical Name or Material D-Mannitol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'