missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Methionine, 99%

Catalog Number p-7025621
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
348-67-4
C5H11NO2S
149.21
MFCD00002622
FFEARJCKVFRZRR-SCSAIBSYSA-N
d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid
84815
CHEBI:16867
(2R)-2-amino-4-methylsulfanylbutanoic acid
CSCC[C@@H](N)C(O)=O
This item is not returnable. View return policy
348-67-4
C5H11NO2S
149.21
MFCD00002622
FFEARJCKVFRZRR-SCSAIBSYSA-N
d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid
84815
CHEBI:16867
(2R)-2-amino-4-methylsulfanylbutanoic acid
CSCC[C@@H](N)C(O)=O

D-Methionine is used to rescue noise-induced hearing loss. It is used to improve the nutritive value of animal feed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Methionine is used to rescue noise-induced hearing loss. It is used to improve the nutritive value of animal feed.

Solubility
Soluble in water. 53 g/L (20°C).

Notes
Store away from oxidizing agents. Store in a cool, dry condition in a well sealed container.
TRUSTED_SUSTAINABILITY
Melting Point ∼280°C (decomposition)
Beilstein 1722293
Merck Index 14,5975
Solubility Information Soluble in water. 53g/L (20°C).
Formula Weight 149.21
Percent Purity 99%
Quantity 5 g
Optical Rotation −23° (c=2 in 2N HCl)
Chemical Name or Material D-Methionine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'