Learn More
D(-)-Phenylglycinamide, 99%, Thermo Scientific Chemicals
£57.60 - £203.00
Chemical Identifiers
CAS | 6485-67-2 |
---|---|
Molecular Formula | C8H10N2O |
Molecular Weight (g/mol) | 150.18 |
MDL Number | MFCD06799064 |
InChI Key | KIYRSYYOVDHSPG-SSDOTTSWSA-N |
Synonym | r-2-amino-2-phenylacetamide, 2r-2-amino-2-phenylacetamide, d--phenylglycinamide, d-phenylglycinamide, unii-2ndy0ko51e, 2r-2-amino-2-phenylethanamide, 2ndy0ko51e, d-phenylglycine amide, d--phenylglycinamide,, d---phenylglycine amide |
PubChem CID | 729987 |
IUPAC Name | (2R)-2-amino-2-phenylacetamide |
SMILES | C1=CC=C(C=C1)C(C(=O)N)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10256101
|
Thermo Scientific Acros
346700050 |
5 g | Glass bottle |
£57.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10307521
|
Thermo Scientific Acros
346700250 |
25 g | Glass bottle |
£203.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6485-67-2 | |
150.18 | |
KIYRSYYOVDHSPG-SSDOTTSWSA-N | |
729987 | |
C1=CC=C(C=C1)C(C(=O)N)N |
C8H10N2O | |
MFCD06799064 | |
r-2-amino-2-phenylacetamide, 2r-2-amino-2-phenylacetamide, d--phenylglycinamide, d-phenylglycinamide, unii-2ndy0ko51e, 2r-2-amino-2-phenylethanamide, 2ndy0ko51e, d-phenylglycine amide, d--phenylglycinamide,, d---phenylglycine amide | |
(2R)-2-amino-2-phenylacetamide |
Specifications
6485-67-2 | |
White to Yellow | |
98.5% min. (GC) | |
C6H5CH(NH2)C(O)NH2 | |
−48.5° (20°C c=0.55,EtOH) | |
r-2-amino-2-phenylacetamide, 2r-2-amino-2-phenylacetamide, d--phenylglycinamide, d-phenylglycinamide, unii-2ndy0ko51e, 2r-2-amino-2-phenylethanamide, 2ndy0ko51e, d-phenylglycine amide, d--phenylglycinamide,, d---phenylglycine amide | |
KIYRSYYOVDHSPG-SSDOTTSWSA-N | |
(2R)-2-amino-2-phenylacetamide | |
729987 | |
99% | |
D(-)-Phenylglycinamide, 99% |
125.0°C to 129.0°C | |
Authentic | |
C8H10N2O | |
MFCD06799064 | |
− 48.50 | |
Solubility in water: soluble. Other solubilities: better soluble ph <6 | |
C1=CC=C(C=C1)C(C(=O)N)N | |
150.18 | |
150.18 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
- D(-)-Phenylglycinamide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only