Learn More
Thermo Scientific Chemicals D-Pinitol, 95%
£29.40 - £338.00
Chemical Identifiers
CAS | 10284-63-6 |
---|---|
Molecular Formula | C7H14O6 |
Molecular Weight (g/mol) | 194.18 |
MDL Number | MFCD00216659 |
InChI Key | DSCFFEYYQKSRSV-FEPQRWDDSA-N |
Synonym | d-pinitol, pinitol, 3-o-methyl-d-chiro-inositol, d-+-pinitol, inzitol, pinit, +-pinitol, methylinositol, sennitol, d-ononitol |
PubChem CID | 164619 |
IUPAC Name | (1S,2S,4S,5R)-6-methoxycyclohexane-1,2,3,4,5-pentol |
SMILES | COC1[C@@H](O)[C@@H](O)C(O)[C@H](O)[C@H]1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15413158
|
Thermo Scientific Chemicals
H56648.MD |
250 mg |
£29.40
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15493148
|
Thermo Scientific Chemicals
H56648.03 |
1 g |
£83.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15403158
|
Thermo Scientific Chemicals
H56648.06 |
5 g |
£338.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is a precursor to biologically active fluorinated isosteres of inositol, which show cell growth inhibitory properties and has shown antidiabetic properties in mice. It is believed to be a salt stress regulator in a wide range of plants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a precursor to biologically active fluorinated isosteres of inositol, which show cell growth inhibitory properties and has shown antidiabetic properties in mice. It is believed to be a salt stress regulator in a wide range of plants.
Solubility
Soluble in water.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Chemical Identifiers
10284-63-6 | |
194.18 | |
DSCFFEYYQKSRSV-FEPQRWDDSA-N | |
164619 | |
COC1[C@@H](O)[C@@H](O)C(O)[C@H](O)[C@H]1O |
C7H14O6 | |
MFCD00216659 | |
d-pinitol, pinitol, 3-o-methyl-d-chiro-inositol, d-+-pinitol, inzitol, pinit, +-pinitol, methylinositol, sennitol, d-ononitol | |
(1S,2S,4S,5R)-6-methoxycyclohexane-1,2,3,4,5-pentol |
Specifications
10284-63-6 | |
C7H14O6 | |
14,10361 | |
Soluble in water. | |
+60° (c=1 in Water) | |
(1S,2S,4S,5R)-6-methoxycyclohexane-1,2,3,4,5-pentol | |
164619 | |
95% |
178°C to 185°C | |
MFCD00216659 | |
d-pinitol, pinitol, 3-o-methyl-d-chiro-inositol, d-+-pinitol, inzitol, pinit, +-pinitol, methylinositol, sennitol, d-ononitol | |
DSCFFEYYQKSRSV-FEPQRWDDSA-N | |
COC1[C@@H](O)[C@@H](O)C(O)[C@H](O)[C@H]1O | |
194.18 | |
194.18 | |
D-Pinitol |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only