missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(-)-Prolinamide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It can be used in the synthesis of proline derivatives as efficient organocatalysts for aldol reaction on water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used in the synthesis of proline derivatives as efficient organocatalysts for aldol reaction on water.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It can be used in the synthesis of proline derivatives as efficient organocatalysts for aldol reaction on water.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 100°C to 103°C |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Optical Rotation | +107° (c=2 in Ethanol) |
Formula Weight | 114.15 |
Percent Purity | 98% |
Chemical Name or Material | D-(-)-Prolinamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'