missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Proline methyl ester hydrochloride, 98%, Thermo Scientific™
£46.80 - £163.00
Chemical Identifiers
CAS | 65365-28-8 |
---|---|
Molecular Formula | C6H11NO2·ClH |
Molecular Weight (g/mol) | 165.62 |
InChI Key | HQEIPVHJHZTMDP-NUBCRITNSA-N |
Synonym | d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride |
PubChem CID | 44629857 |
IUPAC Name | methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride |
SMILES | COC(=O)C1CCCN1.Cl |
Description
Chemical Identifiers
65365-28-8 | |
165.62 | |
d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride | |
methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride |
C6H11NO2·ClH | |
HQEIPVHJHZTMDP-NUBCRITNSA-N | |
44629857 | |
COC(=O)C1CCCN1.Cl |
Specifications
65365-28-8 | |
White | |
97.5 to max. 102.5% min. (Argentometry) | |
C6H11NO2·ClH | |
d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride | |
COC(=O)C1CCCN1.Cl | |
165.62 | |
165.62 | |
Crystalline Powder or Powder |
42°C to 49°C | |
Authentic | |
Glass bottle | |
35 | |
HQEIPVHJHZTMDP-NUBCRITNSA-N | |
methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride | |
44629857 | |
98% | |
D-Proline methyl ester hydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- D-Proline methyl ester hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only