missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Quinic acid, 98+%, Thermo Scientific Chemicals

Product Code. 10659014
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-95-2
Molecular Formula C7H12O6
Molecular Weight (g/mol) 192.17
MDL Number MFCD00003864
InChI Key AAWZDTNXLSGCEK-UHFFFAOYNA-N
Synonym quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi
PubChem CID 6508
IUPAC Name (3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
SMILES OC1CC(O)(CC(O)C1O)C(O)=O

Specifications

Melting Point 166.0°C to 168.0°C
Color White to Yellow
Ignition Residue 0.1% max.
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (HO)4C6H7CO2H
Quantity 100 g
Merck Index 15,8175
Specific Rotation Condition - 44.00 (20.00°C c=11.2,H2O)
Specific Rotation − 44.00
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: soluble in glacial acetic acid and alcohol
Formula Weight 192.17
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-Quinic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'