missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-(+)-Raffinose pentahydrate, 99%
Click to view available options
Quantity:
10 g
50 g
Description
D-(+)-Raffinose pentahydrate is used as a carbon source in yeast culture media. It is also used in moisturizers, prevent skin ageing, promotes skin regeneration, additive in low-cariogenic food and drinks, non-digestible with lower efficiency than lactose or sucrose and stimulates the growth of lactobacilli and bifidobacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-(+)-Raffinose pentahydrate is used as a carbon source in yeast culture media. It is also used in moisturizers, prevent skin ageing, promotes skin regeneration, additive in low-cariogenic food and drinks, non-digestible with lower efficiency than lactose or sucrose and stimulates the growth of lactobacilli and bifidobacteria.
Solubility
Soluble in pyridine and water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
D-(+)-Raffinose pentahydrate is used as a carbon source in yeast culture media. It is also used in moisturizers, prevent skin ageing, promotes skin regeneration, additive in low-cariogenic food and drinks, non-digestible with lower efficiency than lactose or sucrose and stimulates the growth of lactobacilli and bifidobacteria.
Solubility
Soluble in pyridine and water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications
Melting Point | 78°C to 82°C |
Assay Percent Range | 0.99 |
Quantity | 10 g |
Beilstein | 4285763 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in pyridine and water. |
Formula Weight | 594.52 (504.45 Anhydrous) |
Chemical Name or Material | D-(+)-Raffinose pentahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'