missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Salicin, 99+%, Thermo Scientific Chemicals

£24.50 - £253.00

Chemical Identifiers

CAS 138-52-3
Molecular Formula C13H18O7
Molecular Weight (g/mol) 286.27
MDL Number MFCD00006590
InChI Key NGFMICBWJRZIBI-UJPOAAIJSA-N
Synonym salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
PubChem CID 439503
ChEBI CHEBI:17814
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
SMILES C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10413775
View Documents Promotion Details
Thermo Scientific Acros
132590050
5 g Glass bottle
£24.50
5g
Please to purchase this item. Need a web account? Register with us today!
10387110
View Documents Promotion Details
Thermo Scientific Acros
132590250
25 g Glass bottle
£77.60
25g
Please to purchase this item. Need a web account? Register with us today!
10397110
View Documents Promotion Details
Thermo Scientific Acros
132591000
100 g Glass bottle
£253.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

138-52-3
286.27
NGFMICBWJRZIBI-UJPOAAIJSA-N
439503
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
C13H18O7
MFCD00006590
salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
CHEBI:17814
C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O

Specifications

138-52-3
White
99% min. (HPLC)
MFCD00006590
15, 8460
-61.5
Solubility in water: 36g/L (15°C),250g/L (60°C). Other solubilities: insoluble in ether
C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O
286.27
CHEBI:17814
99+%
D(-)-Salicin, 99+%
197°C to 200°C
Authentic
C13H18O7
17, V,7, 113
−61.50° (20°C c=5,water)
salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
NGFMICBWJRZIBI-UJPOAAIJSA-N
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
439503
286.27
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.

GHS P Statement
Wear eye protection/face protection.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 205-331-6

missing translation for 'rtecsNumber' : LZ5901700

missing translation for 'tsca' : TSCA

Product Identifier
  • D(-)-Salicin

Signal Word
  • Warning

Hazard Category
  • Respiratory sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only