missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Sorbitol, 97%

Product Code. 10113570 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Unit Size:
100g
10kg
1kg
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10113570

Brand: Thermo Scientific Chemicals 132730025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-70-4 | C6H14O6 | 182.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00004708
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
PubChem CID 5780
ChEBI CHEBI:17924
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Melting Point 95.0°C to 99.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on anhydrous substance) (HPLC)
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 1533
Merck Index 158851
Specific Rotation Condition + 4.00 (20.00°C per eur. pharm.)
Specific Rotation + 4.00
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in acetic acid, soluble in glycerol and propylene glycol, slightly soluble in methanol, ethanol, phenol,, acetamide, acetone, dmf and pyridine, practically insoluble in most other org. sol
Formula Weight 182.17
Percent Purity 97%
Physical Form Granular Crystalline Powder
Chemical Name or Material D-Sorbitol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.