missing translation for 'onlineSavingsMsg'
Learn More

D-Sorbitol hexaacetate, 97%, Thermo Scientific™

Product Code. 15447369
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7208-47-1
Molecular Formula C18H26O12
Molecular Weight (g/mol) 434.394
MDL Number MFCD00008715
InChI Key NJVBTKVPPOFGAT-XMTFNYHQSA-N
Synonym d-sorbitol hexaacetate, sorbitol hexaacetate, sorbitol, acetylated, 2r,3r,4r,5s-1,2,4,5,6-pentakis acetyloxy hexan-3-yl acetate
PubChem CID 9802952
IUPAC Name [(2S,3R,4R,5R)-2,3,4,5,6-pentaacetyloxyhexyl] acetate
SMILES CC(=O)OCC(C(C(C(COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 100°C to 104°C
Color White
Quantity 100 g
Formula Weight 434.39
Percent Purity 97%
Physical Form Powder
Chemical Name or Material D-Sorbitol hexaacetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'