missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D(+)-Sucrose, 99.7%, for biochemistry Product Code.: 10134050

Thermo Scientific Chemicals D(+)-Sucrose, 99.7%, for biochemistry

Product Code. 10134050
1 kg, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10134050

Brand: Thermo Scientific Chemicals 177140010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D(+)-Sucrose, CAS # 57-50-1, is a disaccharide used in a variety of biochemical applications such as protein purification. It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • D(+)-Sucrose is a disaccharide carbohydrate
  • It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose

Applications

  • D(+)-Sucrose can be used for protein and nucleic acid purification
  • Its fatty acid esters have laboratory research applications in transdermal systems
  • It is also used for sucrose-gradient centrifugation, which may be employed for the selective isolation of Nocardia species from soil
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 190.0°C to 192.0°C
Color Colorless
pH 6.5 to 7.5 (10% soln.)
Loss on Drying 0.1 % max.
Infrared Spectrum Authentic
Assay Percent Range 99.7% min. (HPLC)
Packaging Plastic bottle
Quantity 1 kg
Beilstein 31424
Merck Index 159012
Specific Rotation (20°C, 589nm) (c=26, H2O) > +65.9°
Solubility Information Solubility in water: 1970g/L (15°C). Other solubilities: moderately soluble in glycerol and pyridine
Formula Weight 342.29
Percent Purity 99.7%
Grade Biochemistry
Insoluble Matter 0.0007% max. (in water)
Physical Form Liquid
Chemical Name or Material D(+)-Sucrose
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463305 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog Number177140250,177140010,177142500,177140050
Lot NumberA0453886 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number177140250,177140010,177142500,177140050
Lot NumberA0453885 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number177140250,177140010,177142500,177140050
Lot NumberA0447978 Certificate TypeCertificate of Analysis Date10/04/2023 Catalog Number177140250,177140010,177142500,177140050
Lot NumberA0420137 Certificate TypeCertificate of Analysis Date23/12/2022 Catalog Number177140250,177140010,177142500,177140050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D(+)-Sucrose, 99.7%, for biochemistry >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.