missing translation for 'onlineSavingsMsg'
Learn More
D(-)-Tartaric Acid (unnatural Tartaric Acid) 99%, Thermo Scientific Chemicals Product Code.: 10306930

D(-)-Tartaric Acid (unnatural Tartaric Acid) 99%, Thermo Scientific Chemicals

Product Code. 10306930
25 g, Glass Bottle
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10306930

Brand: Thermo Scientific Acros 137860250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-71-7
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00004238
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym d-tartaric acid, d---tartaric acid, 2s,3s-2,3-dihydroxysuccinic acid, d--tartaric acid, d-threaric acid, --d-tartaric acid, 2s,3s-2,3-dihydroxybutanedioic acid, --tartaric acid, s,s-tartaric acid, 2s,3s---tartaric acid
PubChem CID 439655
ChEBI CHEBI:15672
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 167°C to 171°C
Color White
Assay Percent Range 99%
Linear Formula HO2CCH(OH)CH(OH)CO2H
Beilstein 03, 520
Fieser 16,304
Merck Index 15, 9203
IUPAC Name (2S,3S)-2,3-dihydroxybutanedioic acid
Formula Weight 150.09
Percent Purity ≥98.5%
Quantity 25 g
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (1 g, 105°C, 3 hrs)
Packaging Glass Bottle
Physical Form Crystals or Crystalline Powder
Chemical Name or Material D(-)-Tartaric acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • D(-)-Tartaric acid (unnatural tartaric acid)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : D(-)-Tartaric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D(-)-Tartaric Acid (unnatural Tartaric Acid) 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.