missing translation for 'onlineSavingsMsg'
Learn More

D-tert-Leucine, 99%, Thermo Scientific Chemicals

Product Code. 11378965
Click to view available options
Quantity:
100 mg
500 mg
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

D-tert-Leucine is used for synthesis of amino acids e.g. in the synthesis of aldehydes by double alkylation of unsaturated aldimines of tert-Leu esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-tert-Leucine is used for synthesis of amino acids e.g. in the synthesis of aldehydes by double alkylation of unsaturated aldimines of tert-Leu esters.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26782-71-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00004265
InChI Key NPDBDJFLKKQMCM-UHFFFAOYNA-N
Synonym d-tert-leucine, h-d-tle-oh, d-tert-butylglycine, r-2-amino-3,3-dimethylbutanoic acid, d-t-leucine, r-tert-leucine, 2r-2-amino-3,3-dimethylbutanoic acid, d-t-butylglycine, h-tbu-d-gly-oh, d-valine, 3-methyl
PubChem CID 6950340
IUPAC Name (2R)-2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)C(N)C(O)=O

Specifications

Melting Point >300°C
Beilstein 1721825
Solubility Information Soluble in water.
Formula Weight 131.18
Percent Purity 99%
Quantity 500 mg
Optical Rotation −7° (c=5 in 5N HCl)
Chemical Name or Material D-tert-Leucine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'