missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Tryptophan, 99%

Product Code. 10533341 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10533341

Brand: Thermo Scientific Chemicals 168630250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 153-94-6 | C11H12N2O2 | 204.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153-94-6
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00005647
InChI Key QIVBCDIJIAJPQS-SECBINFHSA-N
Synonym d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh
PubChem CID 9060
ChEBI CHEBI:16296
IUPAC Name (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 282.0°C to 285.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Loss on Drying 0.5% max.
Packaging Glass Bottle
Specific Rotation + 31.50
Solubility Information Solubility in water: 11g/L (20°C). Other solubilities: soluble in alkali hydroxides, soluble in hot alcohol, insoluble in chloroform
Specific Rotation Condition + 31.50 (24.00°C c=1,H2O)
Quantity 25 g
Formula Weight 204.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material D(+)-Tryptophan
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.