missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-Tryptophan, 99% Product Code.: 11428357

Thermo Scientific Chemicals D-Tryptophan, 99%

Product Code. 11428357
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11428357

Brand: Thermo Scientific Chemicals A18426.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-Tryptophan is used for human embryonic kidney cell (HEK-293, ATCC: CRL-1573) culture. The product is a sweetener used to study the release of incretins from enteroendocrine cells triggered by sugar but not by sweeteners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tryptophan is used for human embryonic kidney cell (HEK-293, ATCC: CRL-1573) culture. The product is a sweetener used to study the release of incretins from enteroendocrine cells triggered by sugar but not by sweeteners.

Solubility
Soluble in 0.5M hydrochloric acid (50 mg/ml), water (11 mg/ml at 20°C), hot alcohol, and dimethyl sulfoxide. Insoluble in chloroform.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153-94-6
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00005647
InChI Key QIVBCDIJIAJPQS-SECBINFHSA-N
Synonym d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-ohShow MoreShow Less
PubChem CID 9060
ChEBI CHEBI:16296
IUPAC Name (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 273°C (decomposition)
Beilstein 86198
Solubility Information Soluble in 0.5M hydrochloric acid (50mg/ml),water (11mg/ml at 20°C),hot alcohol,and dimethyl sulfoxide. Insoluble in chloroform.
Quantity 100 g
Formula Weight 204.23
Optical Rotation +31.5° (c=1 in Water)
Percent Purity 99%
Chemical Name or Material D-Tryptophan

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250809 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA18426.22,A18426.06,A18426.14
Lot Number10243216 Certificate TypeCertificate of Analysis Date14/09/2022 Catalog NumberA18426.22,A18426.06,A18426.14
Lot Number5004A04S Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18426.22,A18426.06,A18426.14
Lot Number10228061 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18426.22,A18426.06,A18426.14
Lot Number5002V19U Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18426.22,A18426.06,A18426.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-Tryptophan, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.