Learn More
D-Tyrosine, 99%, Thermo Scientific Chemicals
£21.60 - £320.00
Chemical Identifiers
CAS | 556-02-5 |
---|---|
Molecular Formula | C9H11NO3 |
Molecular Weight (g/mol) | 181.19 |
MDL Number | MFCD00063073 |
InChI Key | OUYCCCASQSFEME-SVGMAFHSNA-N |
Synonym | d-tyrosine, h-d-tyr-oh, d-tyr, r-2-amino-3-4-hydroxyphenyl propanoic acid, d-tyrosin, 2r-2-amino-3-4-hydroxyphenyl propanoic acid, 3-4-hydroxyphenyl-d-alanine, d-p-tyrosine, tyrosine, d, d-+-tyrosine |
PubChem CID | 71098 |
ChEBI | CHEBI:28479 |
IUPAC Name | (2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid |
SMILES | N[C@H](CC1=CC=C(O)C=C1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11483288
|
Thermo Scientific Alfa Aesar
A17709.03 |
1 g |
£21.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493288
|
Thermo Scientific Alfa Aesar
A17709.06 |
5 g |
£71.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403298
|
Thermo Scientific Alfa Aesar
A17709.14 |
25 g |
£320.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D-Tyrosine is synthesized from phenylalanine. It is also the precursor of epinephrine, thyroid hormones, and melanin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Tyrosine is synthesized from phenylalanine. It is also the precursor of epinephrine, thyroid hormones, and melanin.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
556-02-5 | |
181.19 | |
OUYCCCASQSFEME-SVGMAFHSNA-N | |
71098 | |
(2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid |
C9H11NO3 | |
MFCD00063073 | |
d-tyrosine, h-d-tyr-oh, d-tyr, r-2-amino-3-4-hydroxyphenyl propanoic acid, d-tyrosin, 2r-2-amino-3-4-hydroxyphenyl propanoic acid, 3-4-hydroxyphenyl-d-alanine, d-p-tyrosine, tyrosine, d, d-+-tyrosine | |
CHEBI:28479 | |
N[C@H](CC1=CC=C(O)C=C1)C(O)=O |
Specifications
310°C to 314°C | |
C9H11NO3 | |
2212157 | |
d-tyrosine, h-d-tyr-oh, d-tyr, r-2-amino-3-4-hydroxyphenyl propanoic acid, d-tyrosin, 2r-2-amino-3-4-hydroxyphenyl propanoic acid, 3-4-hydroxyphenyl-d-alanine, d-p-tyrosine, tyrosine, d, d-+-tyrosine | |
OUYCCCASQSFEME-SVGMAFHSNA-N | |
(2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid | |
71098 | |
181.19 | |
+11° (c=4 in 1N HCl) |
556-02-5 | |
MFCD00063073 | |
14,9839 | |
Soluble in water. | |
N[C@H](CC1=CC=C(O)C=C1)C(O)=O | |
181.19 | |
CHEBI:28479 | |
99% | |
D-Tyrosine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 209-112-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only