Learn More
DAB EASY tablets, for immunohistochemistry, 10 mg/tablet
DAB, or 3,3-diaminobenzidine tetrahydrochloride, EASY tablets for immunohistochemistry, CAS # 7411-49-6, is a chromogenic substrate used to observe peroxidase activity. In immunohistochemistry, DAB is used to catalyze peroxidase-bound antibodies in order to visualize protein presence. | CAS: 7411-49-6
Brand: Thermo Scientific Acros 328005000
190.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- 3,3′-Diaminobenzidine is a derivative of benzene and the precursor to polybenzimidazole
Applications
- DAB is mainly used to visualize peroxidase activity. During this chemical reaction, DAB is a hydrogen donor in the presence of peroxide. When DAB is oxidized, an insoluble brown end-product is produced.
- DAB has been used for staining and oxidative detection of mitochondria
- It has also been utilized for electron microscopy applications

Specifications
DAB EASY tablets | |
7411-49-6, 10043-35-3 | |
Solubility in water: soluble. | |
MFCD00007725 |
Tablets | |
Plastic bottle | |
500 mg |
Safety and Handling
- DAB EASY tablets
Signal Word
- Danger
Hazard Category
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Boric acid (H3BO3),3,3'-Diaminobenzidine tetrahydrochloride
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466977 | Certificate of Analysis | 01/11/2024 | 328005000 |
A0458496 | Certificate of Analysis | 15/12/2023 | 328005000 |
A0425111 | Certificate of Analysis | 25/08/2023 | 328005000 |
A0453720 | Certificate of Analysis | 25/08/2023 | 328005000 |
A0411159 | Certificate of Analysis | 13/12/2022 | 328005000 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only