missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ DACM, N-(7-Dimethylamino-4-Methylcoumarin-3-yl))Maleimide
N-(7-dimethylamino-4-methylcoumarin-3-yl)maleimide (DACM)
Brand: Molecular Probes™ D10251
276.64 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
The thiol reactive coumarin, DACM can be used to create blue-fluorescent bioconjugates. When compared with AMCA conjugates, conjugates of the UV light-excitable 7-dialkylaminocoumarin fluorophore have slightly longer-wavelength emission spectra (∼470 nm). The unreacted reagent is nonfluorescent and can also be used to quantitate free thiols.Specifications
10 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Methylcoumarin | |
470 |
Maleimide | |
Thiol | |
Maleimide | |
Classic Dyes |
For Research Use Only. Not for use in diagnostic procedures.