missing translation for 'onlineSavingsMsg'
Learn More

Melford Daidzein

Product Code. p-200087238
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30018226

Brand: Melford D200100.025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inactive analog of Genistein.

Inactive analog of Genistein. Inhibits casein kinase II activity.

Specifications

CAS 486-66-8
Molecular Formula C15H10O4
Quantity 25 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.