Learn More
Daidzein, 98%, Thermo Scientific Chemicals
£61.50 - £208.00
Chemical Identifiers
CAS | 486-66-8 |
---|---|
Molecular Formula | C15H10O4 |
Molecular Weight (g/mol) | 254.24 |
MDL Number | MFCD00016954 |
InChI Key | ZQSIJRDFPHDXIC-UHFFFAOYSA-N |
Synonym | daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone |
PubChem CID | 5281708 |
ChEBI | CHEBI:28197 |
IUPAC Name | 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one |
SMILES | C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10613552
|
Thermo Scientific Acros
328230250 |
25 mg | Glass bottle |
£61.50
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10209613
|
Thermo Scientific Acros
328231000 |
100 mg | Glass bottle |
£208.00
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
486-66-8 | |
254.24 | |
ZQSIJRDFPHDXIC-UHFFFAOYSA-N | |
5281708 | |
7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one |
C15H10O4 | |
MFCD00016954 | |
daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone | |
CHEBI:28197 | |
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O |
Specifications
486-66-8 | |
Authentic | |
C15H10O4 | |
18,II,1 | |
daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone | |
ZQSIJRDFPHDXIC-UHFFFAOYSA-N | |
7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one | |
5281708 | |
254.24 | |
Powder |
White | |
98% | |
MFCD00016954 | |
15, 2798 | |
Solubility in water: insoluble. Other solubilities: soluble in dmso | |
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O | |
254.24 | |
CHEBI:28197 | |
98% | |
Daidzein |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-635-4
- Daidzein
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only