missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva DALT-Twelve Gel Caster
Brand: Cytiva 80-6467-22
Code : GE
Additional Details : Weight : 8.50000kg
Description
Hoefer DALT is an integrated system for reproducible high resolution, high throughput 2-D electrophoresis. The instrument and accessory modules are especially designed for multiple large format acrylamide gels. For added flexibility, DALTtwelve Gel Caster is available for casting up to 14 gels simultaneously. Regular and low-fluorescence glass plates are available for use with the DALT Gel Casting Cassette. Low-fluorescence glass plates ensure high sensitivity when using fluorescent dyes. Gradient gels can be prepared using DALT Gradient Maker.Specifications
DALT-Twelve Gel Caster | |
Ettan DALTtwelve Large Vertical System | |
1 to 14 Gels (1.0 mm thick gels); 1 to 13 Gels (1.5 mm thick gels) |
Acrylamide solution volume (total) 950 mL for 1.0 mm thick gels; 1200 mL for 1.5 mm thick gel | |
20 x 26 x 0.1 cm or 20 x 26 x 0.15 cm |