missing translation for 'onlineSavingsMsg'
Learn More

Manutan Danger biohazard sign, photoluminescent plastic, 250mm x 200mm

Brand:  Manutan A347120

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12806388

  • £15.60 / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

Ideal for low-light areas, warn of immediately hazardous situations.
  • Photoluminescent rigid plastic
  • Dimensions (H x W) : 250mm x 200mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers