Learn More
Molecular Probes™ Dansyl Ethylenediamine, 5-Dimethylaminonaphthalene-1-(N-(2-Aminoethyl))sulfonamide
5-dimethylaminonaphthalene-1-(N-(2-aminoethyl))sulfonamide (dansyl ethylenediamine)
Brand: Molecular Probes™ D112
This item is not returnable.
View return policy
Description
The primary aliphatic amine of dansyl ethylenediamine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247). This fluorophore is lipophilic and environmentally-sensitive.Specifications
100 mg | |
Store at room temperature and protect from light. | |
Room Temperature | |
Dansyl |
Dansyl Ethylenediamine | |
Carboxylic Acid, Ketone, Aldehyde | |
Amine, Ethylenediamine | |
Classic Dyes |
For Research Use Only. Not for use in diagnostic procedures.