Learn More
Tocris Bioscience™ DAPI
Fluorescent DNA stain
Brand:  Tocris Bioscience™ 5748/50
Additional Details : CAS Number : 28718-90-3 Weight : 0.00970kg
 View more versions of this product
Product Code. 18760298
  • £332.60 / 50mg
Estimated Shipment: 08-07-2024
to see stock.
Description
Description
DAPI is a fluorescent DNA stain/ dye. DAPI stain binds to AT-rich regions of DNA and fluorescence increases by approximately 20-fold when bound to double-stranded DNA. DAPI stain can be used in flow cytometry, DNA visualization and quantification in histochemistry/biochemistry, to assess apoptosis, and to visualize mouse embryos or fetal organs. DAPI stain can be used in fixed cells and is cell-permeable at high concentrations providing effective cell staining. It is reported to be more photostable than Hoechst 33342 (Cat. No. 5117). Em/Ex λ = 350/470 nm respectively.
Specifications
Specifications
2-[4-(Aminoiminomethyl)phenyl]-1 H-Indole-6-carboximidamide hydrochloride
50 mg
C16H15N50.2 HCl
0.95
28718-90-3
Fluorescent DNA Stains
350.25