Learn More
Tocris Bioscience™ DAPT
DAPT synthesized to Ancillary Material Grade
Brand: Tocris Bioscience™ TB2634-RMU/10
Additional Details : Weight : 0.00970kg
Description
DAPT is an ancillary Material Grade version of DAPT (Cat. No. 2634). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.Find out more about our Ancillary Material Grade product range.
Specifications
(2 S)-N-[(3,5-Difluorophenyl)acetyl]-L-alanyl-2-phenylglycine 1,1-dimethylethyl ester | |
10 mg | |
C23H26F2N2O4 |
208255-80-5 | |
Secretase Inhibitors | |
0.99 |